siRNA Details
Gene Symbol: Yipf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026417.4 | NP_080693.2 | 1, 2 | 310 | ||
GenBank mRNA | AK009618.1 | - | 1, 2 | 296 | ||
AK017721.1 | - | 1, 2 | 288 | |||
AK017722.1 | - | 1, 2 | 288 | |||
AK083301.1 | - | 1, 2 | 290 | |||
AK159227.1 | - | 1, 2 | 271 | |||
AK166347.1 | - | 1, 2 | 298 | |||
BY709789.1 | - | 1, 2 | 310 |
