siRNA Details
Gene Symbol: RIOK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159749.1 | NP_001153221.1 | 8 | 1372 | ||
NM_018343.2 | NP_060813.2 | 8 | 1372 | |||
XM_017009628.1 | XP_016865117.1 | 6 | 997 | |||
GenBank mRNA | AK001697.1 | - | 1 | 349 | ||
AK002021.1 | - | 8 | 1311 | |||
AK225348.1 | - | 8 | 1293 | |||
BC000953.2 | - | 8 | 1300 | 3449997 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
