siRNA Details
Gene Symbol: Trem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272078.1 | NP_001259007.1 | 5 | 1050 | ||
NM_031254.3 | NP_112544.1 | 5 | 995 | |||
GenBank mRNA | AK039477.1 | - | 5 | 900 | ||
AK155163.1 | - | 5 | 964 | |||
AK171358.1 | - | 2818 | ||||
AY024348.1 | - | 5 | 915 | |||
BC032959.1 | - | 5 | 884 | 1348066 | ||
BC033485.1 | - | 5 | 906 | 4953284 | ||
BC052784.1 | - | 5 | 1004 | 5057491 | ||
BY541740.1 | - | 2 | 281 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
