siRNA Details
Gene Symbol: Hamp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032541.1 | NP_115930.1 | 3 | 206 | ||
GenBank mRNA | AF297664.1 | - | 3 | 206 | ||
AF503444.1 | - | 3 | 206 | |||
BC021587.1 | - | 3 | 189 | 5097807 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
