siRNA Details
Gene Symbol: Lmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA050915.1 | - | 2 | 296 | ||
AK133586.1 | - | 2 | 185 | |||
AW494084.1 | - | 2 | 379 | |||
BC053074.1 | - | 2 | 435 | 5693266 | ||
DV662082.1 | - | 2 | 646 |

More Information
Other Species Targeted

