siRNA Details
Gene Symbol: Trrap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081362.1 | NP_001074831.1 | 71 | 11939 | ||
GenBank mRNA | AK029388.1 | - | 5 | 1348 | ||
AK135094.1 | - | 2 | 2560 | |||
AK147241.1 | - | 71 | 11939 | |||
AK147613.1 | - | 48 | 8455 | |||
AK165003.1 | - | 12 | 2544 | |||
AK167768.1 | - | 6 | 1463 | |||
BC008264.1 | - | 1 | 240 | |||
BC024064.1 | - | 9 | 2112 | |||
BC029023.1 | - | 24 | 4687 | |||
BC092382.1 | - | 2 | 333 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
