siRNA Details
Gene Symbol: AY074887 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145229.2 | NP_660264.1 | 1 | 254 | ||
GenBank mRNA | AY074887.2 | - | 1 | 338 | ||
BC119353.1 | - | 1 | 254 | 8734102 | ||
BC120606.1 | - | 1 | 254 | 40129529 |
