siRNA Details
Gene Symbol: Olfr930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146272.1 | NP_666384.1 | 1 | 199 | ||
GenBank mRNA | BC119252.1 | - | 1 | 281 | 8734001 | |
BC119254.1 | - | 1 | 283 | 8734003 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
