siRNA Details
Gene Symbol: Olfr193 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011791.1 | NP_001011791.1 | 1 | 202 | ||
GenBank mRNA | BC147133.1 | - | 1 | 227 | 9056183 | |
BC147134.1 | - | 1 | 225 | 9056182 |
Gene Symbol: Olfr196 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146779.2 | NP_666990.2 | 1 | 202 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
