siRNA Details
Gene Symbol: Mapkap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290625.1 | NP_001277554.1 | 12 | 2802 | ||
NM_001290626.1 | NP_001277555.1 | 11 | 2800 | |||
NM_177345.4 | NP_796319.1 | 12 | 3128 | |||
GenBank mRNA | AK132263.1 | - | 12 | 2791 | ||
BC012681.1 | - | 1 | 717 | |||
BC048870.1 | - | 1 | 405 | |||
BC090644.1 | - | 11 | 2702 | 30629263 |

More Information
Other Species Targeted

Gene Ontology Categories:
