siRNA Details
Gene Symbol: Defb34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183035.1 | NP_898856.1 | 2 | 170 | ||
GenBank mRNA | AJ578469.1 | - | 2 | 170 |
Gene Symbol: Defb15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139222.3 | NP_631968.1 | 2 | 198 | ||
GenBank mRNA | AJ437648.1 | - | 2 | 198 |

More Information
Other Species Targeted

Gene Ontology Categories:
