siRNA Details
Gene Symbol: Olfr930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146272.1 | NP_666384.1 | 1 | 202 | ||
GenBank mRNA | BC119252.1 | - | 1 | 284 | 8734001 | |
BC119254.1 | - | 1 | 286 | 8734003 |

More Information
Other Species Targeted

Gene Ontology Categories:
