siRNA Details
Gene Symbol: Zswim8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252081.1 | NP_001239010.1 | 3, 4 | 758 | ||
NM_001252082.1 | NP_001239011.1 | 3, 4 | 758 | |||
NM_027996.3 | NP_082272.1 | 3, 4 | 758 | |||
GenBank mRNA | AK029263.1 | - | 2, 3 | 214 | ||
AK053265.1 | - | 3, 4 | 710 | |||
AK147398.1 | - | 3, 4 | 759 | |||
BC085161.1 | - | 3, 4 | 272 | |||
BC151046.1 | - | 3, 4 | 657 | 9087959 | ||
BC151056.1 | - | 3, 4 | 657 | 9087969 | ||
BC151173.1 | - | 3, 4 | 657 | 9088062 | ||
BC172103.1 | - | 3, 4 | 657 | 9054389 |

More Information
Other Species Targeted

Gene Ontology Categories:
