siRNA Details
Gene Symbol: Rnf213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077880.1 | - | 4 | 467 | ||
AK156571.1 | - | 5 | 600 | |||
AK173199.1 | - | 14 | 3813 | |||
BC038025.1 | - | 6 | 758 | |||
BC119284.1 | - | 3 | 288 |

More Information
Other Species Targeted

