siRNA Details
Gene Symbol: Zswim8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252081.1 | NP_001239010.1 | 5 | 998 | ||
NM_001252082.1 | NP_001239011.1 | 5 | 998 | |||
NM_027996.3 | NP_082272.1 | 5 | 998 | |||
GenBank mRNA | AK029263.1 | - | 4 | 454 | ||
AK053265.1 | - | 5 | 950 | |||
AK147398.1 | - | 5 | 999 | |||
BC085161.1 | - | 5 | 512 | |||
BC151046.1 | - | 5 | 897 | 9087959 | ||
BC151056.1 | - | 5 | 897 | 9087969 | ||
BC151173.1 | - | 5 | 897 | 9088062 | ||
BC172103.1 | - | 5 | 897 | 9054389 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
