siRNA Details
Gene Symbol: Fpr-rs6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177316.2 | NP_796290.2 | 1 | 251 | ||
GenBank mRNA | BC104318.1 | - | 1 | 251 | 40044730 | |
BC104319.1 | - | 1 | 251 | 40044732 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
