siRNA Details
Gene Symbol: Plekhh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181073.3 | NP_851418.2 | 29 | 5396 | ||
GenBank mRNA | AA072901.1 | - | 1 | 181 | ||
AK052692.1 | - | 16 | 3071 | |||
AK077737.1 | - | 1 | 1090 | |||
AK122464.1 | - | 29 | 5365 | |||
AK143387.1 | - | 29 | 5213 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
