siRNA Details
Gene Symbol: Krt6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008476.3 | NP_032502.3 | 3 | 809 | ||
GenBank mRNA | AK009394.1 | - | 3 | 812 | ||
BC064470.1 | - | 3 | 783 | |||
BC080820.1 | - | 3 | 809 | 30141946 |
Gene Symbol: Gm5414 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003670.1 | NP_001003670.1 | 3 | 752 |
Gene Symbol: Gm5478 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC046626.1 | - | 3 | 815 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
