siRNA Details
Gene Symbol: BSG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB072923.1 | - | 4 | 346 | ||
AF548371.1 | - | 5 | 740 | |||
BG480530.1 | - | 3 | 345 | |||
BU501595.1 | - | 520 | ||||
D45131.1 | - | 4 | 361 | |||
L20471.1 | - | 4 | 377 | |||
M87879.1 | - | 4 | 354 | |||
X64364.1 | - | 4 | 389 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
