siRNA Details
Gene Symbol: Naip6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010871.2 | NP_035001.2 | 3 | 787 | ||
GenBank mRNA | AF135494.1 | - | 910 | |||
AF381772.1 | - | 3 | 787 | |||
BC116347.2 | - | 3 | 768 | |||
BC141346.1 | - | 3 | 772 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
