siRNA Details
Gene Symbol: Nnat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010923.3 | NP_035053.1 | 2, 3 | 265 | ||
GenBank mRNA | AK003004.1 | - | 2, 3 | 233 | ||
AK160626.1 | - | 2, 3 | 233 | |||
BC036984.1 | - | 2, 3 | 223 | 4981517 | ||
X83570.1 | - | 2, 3 | 220 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_053601, FQ213121, CB711943, U08290.
Gene Ontology Categories:
