siRNA Details
Gene Symbol: Try5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003405.4 | NP_001003405.1 | 3 | 308 | ||
GenBank mRNA | AB009661.1 | - | 3 | 308 | ||
AK007843.1 | - | 3 | 307 | |||
AK131898.1 | - | 3 | 307 | |||
AK160484.1 | - | 3 | 307 | |||
BC139220.1 | - | 3 | 393 | 8862242 | ||
BC139223.1 | - | 3 | 393 | 8862245 |
Gene Symbol: Try4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011646.5 | NP_035776.1 | 3 | 306 | ||
GenBank mRNA | AK003064.1 | - | 3 | 310 | ||
AK008660.1 | - | 3 | 307 | |||
AK008667.1 | - | 3 | 307 | |||
BC050120.1 | - | 3 | 287 | |||
BC061135.1 | - | 3 | 306 | 30306436 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
