siRNA Details
Gene Symbol: Psg18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011963.2 | NP_036093.2 | 2 | 304 | ||
GenBank mRNA | AF128236.1 | - | 2 | 172 | ||
BC138857.1 | - | 2 | 238 | 8861879 | ||
BY713630.1 | - | 2 | 306 | |||
CN704107.1 | - | 2 | 290 | |||
M83346.1 | - | 2 | 172 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
