siRNA Details
Gene Symbol: Psg22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004152.2 | NP_001004152.1 | 2 | 301 | ||
GenBank mRNA | BC050099.1 | - | 2 | 201 | 30014305 | |
BC080694.1 | - | 2 | 313 | 30433272 |
Gene Symbol: Psg19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011964.2 | NP_036094.2 | 2 | 307 | ||
GenBank mRNA | AK145915.1 | - | 1 | 1399 | ||
BC099467.1 | - | 2 | 283 | 30713077 | ||
X98111.1 | - | 2 | 307 | |||
X98112.1 | - | 2 | 307 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
