siRNA Details
Gene Symbol: Hist1h2bc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290380.1 | NP_001277309.1 | 1 | 110 | ||
NM_023422.3 | NP_075911.2 | 1 | 110 | |||
GenBank mRNA | AK005191.1 | - | 1 | 105 | ||
AK005407.1 | - | 1 | 105 | |||
AK011516.1 | - | 1 | 112 | |||
BC019673.1 | - | 1 | 93 | 3993954 | ||
BC082774.1 | - | 1 | 93 |
Gene Symbol: Hist1h2be | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177653.1 | NP_001171124.1 | 2 | 270 | ||
NM_001290530.1 | NP_001277459.1 | 1 | 91 | |||
NM_178194.4 | NP_835501.1 | 2 | 296 | |||
GenBank mRNA | AK030546.1 | - | 2 | 298 | ||
AK049948.1 | - | 2 | 271 | |||
BC069889.1 | - | 1 | 84 | 4217814 |
Gene Symbol: Hist1h2bf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178195.2 | NP_835502.1 | 1 | 77 |
Gene Symbol: Hist1h2bg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178196.4 | NP_835503.1 | 1 | 110 | ||
GenBank mRNA | BC060304.1 | - | 1 | 137 | 4217587 |
Gene Symbol: Hist1h2bh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178197.2 | NP_835504.1 | 1 | 169 | ||
GenBank mRNA | AK158416.1 | - | 118 | |||
BC092138.1 | - | 84 | 30613720 | |||
BC132584.1 | - | 1 | 169 | 40130861 | ||
BC132586.1 | - | 1 | 169 | 40130863 |
Gene Symbol: Hist1h2bj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178198.2 | NP_835505.1 | 1 | 77 | ||
GenBank mRNA | BC115792.1 | - | 69 |
Gene Symbol: Hist1h2bk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175665.2 | NP_783596.1 | 1 | 143 | ||
GenBank mRNA | BC147143.1 | - | 1 | 143 | 9056184 | |
BC147144.1 | - | 1 | 143 | 9056197 |
Gene Symbol: Hist1h2bl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178199.2 | NP_835506.1 | 1 | 77 |
Gene Symbol: Hist1h2bn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178201.2 | NP_835508.1 | 1 | 77 |
Gene Symbol: Hist1h2bq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097979.2 | NP_001091448.2 | 1 | 105 |
Gene Symbol: Hist1h2br | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110555.1 | NP_001104025.1 | 1 | 105 | ||
GenBank mRNA | AK036389.1 | - | 1 | 106 | ||
BC011440.1 | - | 1 | 90 | 3989862 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106114, NM_001109400, NM_001108414, XM_008771682, XM_006254120, and others.
Gene Ontology Categories:
