siRNA Details
Gene Symbol: Krt42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212483.2 | NP_997648.2 | 6 | 1002 | ||
GenBank mRNA | AK019115.1 | - | 3 | 518 | ||
AK019133.1 | - | 3 | 464 | |||
AY459292.1 | - | 6 | 1002 | |||
BC112371.1 | - | 6 | 994 | |||
BC148204.1 | - | 6 | 1002 | 40056537 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
