siRNA Details
Gene Symbol: Ercc6l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013608.2 | NP_001013626.2 | 2 | 228 | ||
NM_023507.3 | NP_075996.2 | 2 | 214 | |||
GenBank mRNA | AF217319.1 | - | 1 | 31 | ||
AK002307.1 | - | 2 | 215 | |||
AK049874.1 | - | 2 | 540 | |||
AK050437.1 | - | 1 | 64 | |||
BU148612.1 | - | 2 | 246 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
