siRNA Details
Gene Symbol: Nat9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025400.3 | NP_079676.1 | 2, 3 | 295 | ||
GenBank mRNA | AK003948.1 | - | 2, 3 | 249 | ||
AK009262.1 | - | 1, 2 | 25 | |||
AK020083.1 | - | 2, 3 | 275 | |||
AK082469.1 | - | 2, 3 | 235 | |||
AK186542.1 | - | 2, 3 | 135 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
