siRNA Details
Gene Symbol: CARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014437.2 | NP_001014437.1 | 3, 4 | 451 | ||
NM_001194997.1 | NP_001181926.1 | 3, 4 | 451 | |||
NM_001751.5 | NP_001742.1 | 2, 3 | 202 | |||
NM_139273.3 | NP_644802.1 | 2, 3 | 202 | |||
XM_017018392.1 | XP_016873881.1 | 2, 3 | 387 | |||
GenBank mRNA | AF288206.1 | - | 2, 3 | 135 | ||
AF288207.1 | - | 2, 3 | 135 | |||
AK222614.1 | - | 2, 3 | 143 | |||
AK289432.1 | - | 2, 3 | 175 | |||
AK295371.1 | - | 2, 3 | 93 | |||
AK296763.1 | - | 3, 4 | 316 | |||
AK298514.1 | - | 3, 4 | 418 | |||
AK302644.1 | - | 3, 4 | 418 | |||
AK308005.1 | - | 3, 4 | 371 | |||
AU133264.1 | - | 2, 3 | 192 | |||
BC002880.2 | - | 2, 3 | 188 | 3940167 | ||
BT009913.1 | - | 2, 3 | 118 | |||
BX647906.1 | - | 3, 4 | 425 | |||
DC376478.1 | - | 2, 3 | 202 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Matching Taqman Gene Expression Assay:
Gene Ontology Categories:
