siRNA Details
Gene Symbol: 1700120K04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007016.1 | - | 1 | 184 | ||
AK007155.1 | - | 1 | 184 | |||
AK018960.1 | - | 1 | 192 | |||
AK018969.1 | - | 1 | 191 | |||
BC049550.1 | - | 176 |
