siRNA Details
Gene Symbol: CBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001757.3 | NP_001748.1 | 2, 3 | 597 | ||
GenBank mRNA | AB124848.1 | - | 2, 3 | 597 | ||
AK314879.1 | - | 2, 3 | 495 | |||
AL548852.3 | - | 2, 3 | 534 | |||
BC002511.2 | - | 2, 3 | 482 | 3050837 | ||
BC015640.2 | - | 2, 3 | 496 | 4642240 | ||
BT019843.1 | - | 2, 3 | 405 | |||
CR541708.1 | - | 2, 3 | 405 | |||
J04056.1 | - | 2, 3 | 498 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
