siRNA Details
Gene Symbol: Pibf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029320.3 | NP_083596.2 | 13 | 2443 | ||
GenBank mRNA | AK006060.1 | - | 4 | 417 | ||
AK017117.1 | - | 4 | 509 | |||
AK045133.1 | - | 13 | 1940 | |||
AK149007.1 | - | 8 | 996 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
