siRNA Details
Gene Symbol: Npcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY394488.1 | - | 5 | 353 | ||
AY394489.1 | - | 6 | 644 | |||
AY395907.1 | - | 8 | 1253 |
Gene Symbol: Nptxr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF316612.1 | - | 2 | 797 | ||
AY394487.1 | - | 3 | 523 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
