siRNA Details
Gene Symbol: Gm4736 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053251.1 | NP_444481.1 | 150 | |||
GenBank mRNA | M11897.1 | - | 125 |
Gene Symbol: Gm8882 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177588.1 | NP_001171059.1 | 3 | 149 | ||
GenBank mRNA | BC171935.1 | - | 3 | 149 | 9054220 | |
BC171936.1 | - | 3 | 144 | 9054221 |
Gene Symbol: Prpmp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024705.2 | NP_001019876.2 | 3 | 136 | ||
GenBank mRNA | AK132583.1 | - | 3 | 151 | ||
BC055856.1 | - | 3 | 127 | 4910526 | ||
BC061494.1 | - | 3 | 127 | |||
BC080748.1 | - | 3 | 127 | |||
BC092075.1 | - | 3 | 131 | 4920522 | ||
BC110664.1 | - | 3 | 134 | 4191067 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
