siRNA Details
Gene Symbol: Zfp932 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145563.2 | NP_663538.2 | 3 | 272 | ||
GenBank mRNA | AK009062.1 | - | 3 | 288 | ||
AK032220.1 | - | 2 | 401 | |||
BC012405.1 | - | 3 | 258 |
Gene Symbol: Gm15446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142681.1 | - | 3 | 618 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
