siRNA Details
Gene Symbol: Olfr47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146370.1 | NP_666482.1 | 1 | 421 | ||
GenBank mRNA | AF102521.1 | - | 1 | 235 | ||
BC120658.1 | - | 1 | 453 | 40129581 | ||
BC120660.1 | - | 1 | 453 | 40129583 |
Gene Symbol: Olfr435 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146653.1 | NP_666864.1 | 1 | 421 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
