siRNA Details
Gene Symbol: Olfr472 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146774.1 | NP_666985.1 | 1 | 139 | ||
GenBank mRNA | BC138079.1 | - | 1 | 231 | 8861098 | |
BC145756.1 | - | 1 | 231 | 40131059 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
