siRNA Details
Gene Symbol: Olfr1416 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147038.1 | NP_667249.1 | 1 | 218 | ||
GenBank mRNA | BC104078.1 | - | 1 | 217 | ||
BC104079.2 | - | 1 | 217 |
Gene Symbol: Olfr1415 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011525.1 | NP_001011525.1 | 1 | 218 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
