siRNA Details
Gene Symbol: Cypt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF463501.1 | - | 1 | 323 | ||
BC147783.1 | - | 1 | 407 | 9087502 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cypt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF463501.1 | - | 1 | 323 | ||
BC147783.1 | - | 1 | 407 | 9087502 |