siRNA Details
Gene Symbol: Pamr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173749.4 | NP_776110.3 | 4, 5 | 668 | ||
GenBank mRNA | AK031254.1 | - | 4, 5 | 540 | ||
AK088017.1 | - | 4, 5 | 567 | |||
BB620582.1 | - | 4, 5 | 535 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
