siRNA Details
Gene Symbol: Glyctk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039586.1 | NP_001034675.1 | 5 | 2180 | ||
NM_174846.4 | NP_777271.3 | 4 | 2316 | |||
GenBank mRNA | AK049425.1 | - | 4 | 2297 | ||
AK052709.1 | - | 4 | 2255 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
