siRNA Details
Gene Symbol: Atp11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293667.1 | NP_001280596.1 | 30 | 6383 | ||
NM_001293668.1 | NP_001280597.1 | 30 | 6364 | |||
NM_015804.3 | NP_056619.1 | 29 | 6279 | |||
GenBank mRNA | AK028779.1 | - | 1 | 1123 | ||
AK078972.1 | - | 1 | 711 | |||
AK147497.1 | - | 30 | 6386 | |||
AK161916.1 | - | 1 | 1306 | |||
BC036322.1 | - | 15 | 4424 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
