siRNA Details
Gene Symbol: Dido1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175551.4 | NP_780760.2 | 16 | 4910 | ||
GenBank mRNA | AK032843.1 | - | 1 | 133 | ||
AY425952.1 | - | 16 | 4812 |

More Information
Other Species Targeted

Gene Ontology Categories:
