siRNA Details
Gene Symbol: Celf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176954.3 | NP_795928.2 | 4 | 427 | ||
GenBank mRNA | AK029796.1 | - | 4 | 1601 | ||
AK083461.1 | - | 3 | 155 | |||
AK132321.1 | - | 4 | 393 | |||
AK158669.1 | - | 3 | 204 | |||
BC079845.1 | - | 4 | 399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
