siRNA Details
Gene Symbol: Agtr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175086.3 | NP_780295.2 | 2 | 294 | ||
GenBank mRNA | AK087228.1 | - | 2 | 294 | ||
BC109152.1 | - | 2 | 241 | |||
BC109153.1 | - | 2 | 241 | 40046982 |
Gene Symbol: Agtr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177322.3 | NP_796296.1 | 3 | 485 | ||
GenBank mRNA | BC036175.1 | - | 3 | 384 | 4989471 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
