siRNA Details
Gene Symbol: Mrpl50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178603.4 | NP_848718.1 | 2 | 1519 | ||
GenBank mRNA | AK002646.1 | - | 2 | 1507 | ||
AK007405.1 | - | 2 | 1516 | |||
AK031229.1 | - | 2 | 1520 | |||
AK049421.1 | - | 2 | 1509 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
