siRNA Details
Gene Symbol: Tanc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181071.3 | NP_851416.2 | 2, 3 | 360 | ||
GenBank mRNA | AK051973.1 | - | 2, 3 | 356 | ||
AK052263.1 | - | 1, 2 | 93 | |||
BP766228.1 | - | 2, 3 | 387 |

More Information
Other Species Targeted

Gene Ontology Categories:
