siRNA Details
Gene Symbol: PLK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004073.3 | NP_004064.2 | 5, 6 | 894 | ||
GenBank mRNA | AB209875.1 | - | 6, 7 | 581 | ||
AJ293866.1 | - | 5, 6 | 894 | |||
BC013899.2 | - | 5, 6 | 755 | 4098112 | ||
U56998.1 | - | 5, 6 | 574 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
