siRNA Details
Gene Symbol: COQ8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020247.4 | NP_064632.2 | 8 | 1242 | ||
XM_005273201.1 | XP_005273258.1 | 8 | 1138 | |||
XM_011544238.1 | XP_011542540.1 | 9 | 1334 | |||
XM_011544239.2 | XP_011542541.1 | 10 | 1357 | |||
XM_011544240.2 | XP_011542542.1 | 8 | 1151 | |||
XM_011544241.2 | XP_011542543.1 | 8 | 1113 | |||
XM_017001852.1 | XP_016857341.1 | 8 | 1332 | |||
GenBank mRNA | AB073905.1 | - | 8 | 1165 | ||
AF218003.1 | - | 1 | 18 | |||
AJ278126.1 | - | 4 | 460 | |||
AK074693.1 | - | 8 | 1185 | |||
AK090494.1 | - | 8 | 979 | |||
AK092784.1 | - | 8 | 965 | |||
AK126200.1 | - | 4 | 1049 | |||
AK297735.1 | - | 5 | 521 | |||
AK301012.1 | - | 10 | 1163 | |||
BC005171.2 | - | 8 | 1191 | 3604948 | ||
BC129930.1 | - | 14 | 2084 | 40010265 | ||
BX648860.1 | - | 8 | 1063 |

More Information
Other Species Targeted

Gene Ontology Categories:
