siRNA Details
Gene Symbol: ULK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322500.1 | NP_001309429.1 | 5 | 961 | ||
NM_001322501.1 | NP_001309430.1 | 5 | 961 | |||
NM_017886.3 | NP_060356.2 | 5 | 961 | |||
NR_136342.1 | 5 | 961 | ||||
XM_011533874.2 | XP_011532176.1 | 5 | 1539 | |||
XM_017006711.1 | XP_016862200.1 | 5 | 1539 | |||
XM_017006712.1 | XP_016862201.1 | 5 | 1536 | |||
GenBank mRNA | AK000581.1 | - | 600 | |||
AK304341.1 | - | 5 | 592 | |||
BC014794.1 | - | 5 | 580 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
